4-(Pentafluorothio)phenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxyphenylsulphur pentafluoride

H33413-03 H33413-06
AAH33413-06EA 454.99 USD
AAH33413-06 AAH33413-03
4-(Pentafluorothio)phenol 97%
4-(Pentafluorothio)phenol
Formula: C₆H₅F₅OS
MW: 220.16
Storage Temperature: Ambient
MDL Number: MFCD03788516
CAS Number: 774-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR