5-Fluoro-2-methylthiobenzamide 97%

Warning

H33362-06 H33362-03
AAH33362-06EA 150.11 USD
AAH33362-06 AAH33362-03
5-Fluoro-2-methylthiobenzamide 97%
5-Fluoro-2-methylthiobenzamide
Formula: C₈H₈FNS
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD09757566
CAS Number: 1250676-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR