2-Fluoro-4-methylsulfonylphenol 97%

Warning

H33358-MD H33358-06 H33358-03
AAH33358-06EA 476.19 USD
AAH33358-06 AAH33358-MD AAH33358-03
2-Fluoro-4-methylsulfonylphenol 97%
2-Fluoro-4-methylsulfonylphenol
Formula: C₇H₇FO₃S
MW: 190.19 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD09264549
CAS Number: 398456-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR