2-Ethoxythiobenzamide 97%

Warning

H33353-03 H33353-MD
AAH33353-MDEA 43.04 USD
AAH33353-MD AAH33353-03
2-Ethoxythiobenzamide 97%
2-Ethoxythiobenzamide
Formula: C₉H₁₁NOS
Melting Pt: 113…119 °C
Storage Temperature: Ambient
MDL Number: MFCD06090837
CAS Number: 725702-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR