N-(4-Chloro-2-pyridyl)thiourea 97%

Warning

H33291-03 H33291-MD
AAH33291-03EA 105.68 USD
AAH33291-03 AAH33291-MD
N-(4-Chloro-2-pyridyl)thiourea 97%
N-(4-Chloro-2-pyridyl)thiourea
Formula: C₆H₆ClN₃S
MW: 187.65
Melting Pt: 182…187 °C
Storage Temperature: Ambient
MDL Number: MFCD22380707
CAS Number: 1824186-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR