5-Fluoro-2-(trifluoromethoxy)aniline 97%

Warning

Synonyms: 5-Fluoro-2-(trifluoromethoxy)benzylamine

H33278-03 H33278-MD
AAH33278-03EA 128.7 USD
AAH33278-03 AAH33278-MD
5-Fluoro-2-(trifluoromethoxy)aniline 97%
5-Fluoro-2-(trifluoromethoxy)aniline
Formula: C₇H₅F₄NO
MW: 195.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652514
CAS Number: 123572-63-4
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR