3-(Ethoxycarbonyl)-2-hydroxy-5,6,7,8-tetrahydro-1-naphthoic acid 96%

Warning

H33253-06 H33253-03
AAH33253-03EA 82.79 USD
AAH33253-03 AAH33253-06
3-(Ethoxycarbonyl)-2-hydroxy-5,6,7,8-tetrahydro-1-naphthoic acid 96%
3-(Ethoxycarbonyl)-2-hydroxy-5,6,7,8-tetrahydro-1-naphthoic acid
Formula: C₁₄H₁₆O₅
MW: 264.28 g/mol
Melting Pt: 172…173 °C
Storage Temperature: Ambient
MDL Number: MFCD09258890
CAS Number: 948006-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR