3-(2,3,4-Trimethoxyphenyl)propionic acid 97%

Warning

Synonyms: 3-(2,3,4-Trimethoxyphenyl)propanoic acid

H33230-03 H33230-14 H33230-06
AAH33230-06EA 104.67 USD
AAH33230-06 AAH33230-03 AAH33230-14
3-(2,3,4-Trimethoxyphenyl)propionic acid 97%
3-(2,3,4-Trimethoxyphenyl)propionic acid
Formula: C₁₂H₁₆O₅
MW: 240.26 g/mol
Melting Pt: 70…73 °C
Storage Temperature: Ambient
EINECS: 251-389-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR