3-(4-Bromophenyl)-1H-pyrazole 97%

H33227-14 H33227-03 H33227-06
AAH33227-06EA 82.89 USD
AAH33227-06 AAH33227-14 AAH33227-03
3-(4-Bromophenyl)-1H-pyrazole 97%
3-(4-Bromophenyl)-1H-pyrazole
Formula: C₉H₇BrN₂
Melting Pt: ca 130 °C (polymorphic)
Storage Temperature: Ambient
MDL Number: MFCD01940434
CAS Number: 73387-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR