3-(1H-Pyrazol-3-yl)pyridine 95%

Warning

H33201-06 H33201-03
AAH33201-06EA 518.37 USD
AAH33201-06 AAH33201-03
3-(1H-Pyrazol-3-yl)pyridine 95%
3-(1H-Pyrazol-3-yl)pyridine
Formula: C₈H₇N₃
MW: 145.16 g/mol
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD03407951
CAS Number: 45887-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR