6-Quinoxalinecarboxylic acid 95%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: Quinoxaline-6-carboxylic acid

H33157-03 H33157-06 H33157-14
AAH33157-03EA 68.6 USD
AAH33157-03 AAH33157-14 AAH33157-06
6-Quinoxalinecarboxylic acid 95%
6-Quinoxalinecarboxylic acid
Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
Melting Pt: 248…251 °C
Storage Temperature: Ambient
MDL Number: MFCD01365834
CAS Number: 6925-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR