4-(4-Chlorophenoxy)-3-(trifluoromethyl)aniline 97%

Warning

H33106-03 H33106-06
AAH33106-03EA 75.52 USD
AAH33106-03 AAH33106-06
4-(4-Chlorophenoxy)-3-(trifluoromethyl)aniline 97%
4-(4-Chlorophenoxy)-3-(trifluoromethyl)aniline
Formula: C₁₃H₉ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD01908501
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR