4-(4-Fluorophenoxy)-2-(trifluoromethyl)aniline 97%

Warning

H33043-06 H33043-03
AAH33043-06EA 345.21 USD
AAH33043-06 AAH33043-03
4-(4-Fluorophenoxy)-2-(trifluoromethyl)aniline 97%
4-(4-Fluorophenoxy)-2-(trifluoromethyl)aniline
Formula: C₁₃H₉F₄NO
Storage Temperature: Ambient
MDL Number: MFCD08687913
CAS Number: 946784-69-6
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR