3-Fluoro-5-(trifluoromethoxy)benzoic acid 97%

Warning

H33011-06 H33011-03
AAH33011-06EA 404.04 USD
AAH33011-06 AAH33011-03
3-Fluoro-5-(trifluoromethoxy)benzoic acid 97%
3-Fluoro-5-(trifluoromethoxy)benzoic acid
Formula: C₈H₄F₄O₃
MW: 224.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18783221
CAS Number: 1242258-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR