2-Fluoro-6-methoxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H32994-MD H32994-03
AAH32994-03EA 228.99 USD
AAH32994-03 AAH32994-MD
2-Fluoro-6-methoxypyridine 97%
2-Fluoro-6-methoxypyridine
Formula: C₆H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD09998161
CAS Number: 116241-61-3
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR