2-Methoxy-3-(trifluoromethyl)benzylamine 97%, JRD

Danger

H32993-03 H32993-MD
AAH32993-MDEA 70.01 USD
AAH32993-MD AAH32993-03
2-Methoxy-3-(trifluoromethyl)benzylamine 97%, JRD
2-Methoxy-3-(trifluoromethyl)benzylamine
Formula: C₉H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09832318
CAS Number: 1017778-87-8
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR