3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

H32975-06 H32975-14
AAH32975-14EA 234.21 USD
AAH32975-14 AAH32975-06
3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride 98%
3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride
Formula: C₈H₇ClF₆N₂
MW: 280.60 g/mol
Melting Pt: ca 208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03094165
CAS Number: 502496-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR