4'-Fluoro-3'-hydroxyacetophenone 98%

Warning

H32965-03 H32965-MD
AAH32965-03EA 541.82 USD
AAH32965-03 AAH32965-MD
4'-Fluoro-3'-hydroxyacetophenone 98%
4'-Fluoro-3'-hydroxyacetophenone
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 119…126 °C
Storage Temperature: Ambient
MDL Number: MFCD12545938
CAS Number: 949159-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR