3-Methyl-5-(trifluoromethoxy)phenylacetic acid 97%, JRD

Warning

H32942-MD H32942-03
AAH32942-03EA 298.59 USD
AAH32942-03 AAH32942-MD
3-Methyl-5-(trifluoromethoxy)phenylacetic acid 97%, JRD
3-Methyl-5-(trifluoromethoxy)phenylacetic acid
Formula: C₉H₉F₃O₃
Boiling Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD08741405
CAS Number: 1000339-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR