3,5-Difluorosalicylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Difluoro-2-hydroxybenzoic acid

H32940-MD H32940-03
AAH32940-03EA 137.99 USD
AAH32940-03 AAH32940-MD
3,5-Difluorosalicylic acid 98+%
3,5-Difluorosalicylic acid
Formula: C₇H₄F₂O₃
MW: 174.10
Melting Pt: 187…191 °C
Storage Temperature: Ambient
MDL Number: MFCD06203609
CAS Number: 84376-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR