4-(Methylsulfonyl)phenol 97%

Warning

H32935-09 H32935-03
AAH32935-09EA 228.7 USD
AAH32935-09 AAH32935-03
4-(Methylsulfonyl)phenol 97%
4-(Methylsulphonyl)phenol
Formula: C₇H₈O₃S
MW: 172.20 g/mol
Melting Pt: 90…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00047806
CAS Number: 14763-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR