Methyl-1H-indazole-6-carboxylate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Indazole-6-carboxylic acid methyll esterMethyl 1H-indazole-6-carboxylateMethyl Indazole-6-carboxylate

H32933-03 H32933-MD
AAH32933-MDEA 102.96 USD
AAH32933-MD AAH32933-03
Methyl-1H-indazole-6-carboxylate 95%
Methyl 1H-indazole-6-carboxylate
Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD07371612
CAS Number: 170487-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR