2-(N-Benzyl-N-methylamino)-5-bromopyridine 95%

Warning

H32929-03 H32929-MD
AAH32929-MDEA 108.69 USD
AAH32929-MD AAH32929-03
2-(N-Benzyl-N-methylamino)-5-bromopyridine 95%
2-(N-Benzyl-N-methylamino)-5-bromopyridine
Formula: C₁₃H₁₃BrN₂
Storage Temperature: Ambient
MDL Number: MFCD12913988
CAS Number: 1187386-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR