3-Bromo-5-(trifluoromethoxy)phenylacetonitrile 97%, JRD

Danger

H32908-MD H32908-03
AAH32908-MDEA 46.34 USD
AAH32908-MD AAH32908-03
3-Bromo-5-(trifluoromethoxy)phenylacetonitrile 97%, JRD
3-Bromo-5-(trifluoromethoxy)phenylacetonitrile
Formula: C₉H₅BrF₃NO
MW: 280.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519377
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR