6-Fluoro-3-formylchromone 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 6-Fluorochromone-3-carboxaldehyde6-Fluoro-4-oxo-4H-chromene-3-carbaldehyde

H32857-03
AAH32857-03EA 26.3 USD
AAH32857-03
6-Fluoro-3-formylchromone 97%
6-Fluoro-3-formylchromone
Formula: C₁₀H₅FO₃
MW: 192.15 g/mol
Melting Pt: 155…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00139060
CAS Number: 69155-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR