3'-Fluoro-2'-hydroxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32854-MD
H32854-03
AAH32854-MDEA
225.99
USD
AAH32854-MD
AAH32854-03
3'-Fluoro-2'-hydroxyacetophenone 98%
3'-Fluoro-2'-hydroxyacetophenone
Formula:
C₈H₇FO₂ MW: 154.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09055140 CAS Number: 699-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...