(2-Methyl-3-(trifluoromethyl)phenyl)acetic acid 97%, JRD

Warning

H32853-03 H32853-MD
AAH32853-03EA 231.99 USD
AAH32853-03 AAH32853-MD
(2-Methyl-3-(trifluoromethyl)phenyl)acetic acid 97%, JRD
(2-Methyl-3-(trifluoromethyl)phenyl)acetic acid
Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD09832279
CAS Number: 1000546-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR