6-Hydroxy-1-tetralone 97%

Supplier: Thermo Scientific Chemicals

Warning

H32840-06
AAH32840-06EA 78 USD
AAH32840-06
6-Hydroxy-1-tetralone 97%
6-Hydroxy-1-tetralone
Formula: C₁₀H₁₀O₂
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00156670
CAS Number: 3470-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR