2-Methoxy-4-(trifluoromethyl)benzylamine 97%, JRD

Danger

H32832-03 H32832-MD
AAH32832-MDEA 68.23 USD
AAH32832-MD AAH32832-03
2-Methoxy-4-(trifluoromethyl)benzylamine 97%, JRD
2-Methoxy-4-(trifluoromethyl)benzylamine
Formula: C₉H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD06213016
CAS Number: 771581-99-8
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR