2-Amino-5-phenyl-1,3,4-thiadiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Phenyl-1,3,4-thiadiazol-2-amine2-Amino-5-phenyl-1,3,4-thiadiazol

H32826-03
AAH32826-03EA 83.09 USD
AAH32826-03
2-Amino-5-phenyl-1,3,4-thiadiazole 97%
2-Amino-5-phenyl-1,3,4-thiadiazole
Formula: C₈H₇N₃S
MW: 177.23 g/mol
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00205278
CAS Number: 2002-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR