2,2,2-Trifluoro-N-(2-pyridylmethyl)acetamide 96%

Warning

H32814-06 H32814-03
AAH32814-03EA 49.84 USD
AAH32814-03 AAH32814-06
2,2,2-Trifluoro-N-(2-pyridylmethyl)acetamide 96%
2,2,2-Trifluoro-N-(2-pyridylmethyl)acetamide
Formula: C₈H₇F₃N₂O
Melting Pt: 73…77 °C
Storage Temperature: Ambient
MDL Number: MFCD03361989
CAS Number: 401575-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR