2,3-Difluoro-4-methoxy-DL-phenylalanine 97%, JRD

H32811-MD H32811-03
AAH32811-MDEA 57.09 USD
AAH32811-MD AAH32811-03
2,3-Difluoro-4-methoxy-DL-phenylalanine 97%, JRD
2,3-Difluoro-4-methoxy-DL-phenylalanine
Formula: C₁₀H₁₁F₂NO₃
Melting Pt: ca 260°C dec. (dec.)
Storage Temperature: Ambient
MDL Number: MFCD09832370
CAS Number: 1256482-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR