2-Chloro-6-methoxybenzothiazole 97%

Warning

Synonyms: 2-Chloro-6-methoxy-1,3-benzothiazole2-Chloro-6-methoxybenzo[d]thiazole

H32770-06 H32770-03
AAH32770-03EA 77.41 USD
AAH32770-03 AAH32770-06
2-Chloro-6-methoxybenzothiazole 97%
2-Chloro-6-methoxybenzothiazole
Formula: C₈H₆ClNOS
MW: 199.66 g/mol
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00671510
CAS Number: 2605-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR