3-Fluoro-2-methylcinnamic acid 97%, JRD

Warning

H32762-03 H32762-MD
AAH32762-03EA 190.92 USD
AAH32762-03 AAH32762-MD
3-Fluoro-2-methylcinnamic acid 97%, JRD
3-Fluoro-2-methylcinnamic acid
Formula: C₁₀H₉FO₂
MW: 180.18 g/mol
Melting Pt: 212…215 °C
Storage Temperature: Ambient
MDL Number: MFCD06205416
CAS Number: 155814-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR