3-Methoxythiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methoxybenzenecarbothioamide3-Methoxybenzothioamide

H32734-03 H32734-06
AAH32734-03EA 66.99 USD
AAH32734-03 AAH32734-06
3-Methoxythiobenzamide 97%
3-Methoxythiobenzamide
Formula: C₈H₉NOS
MW: 167.23 g/mol
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD04627361
CAS Number: 64559-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR