3-Ethoxy-2,6-difluoroaniline 97%, JRD

Danger

H32732-03 H32732-06
AAH32732-03EA 92.35 USD
AAH32732-03 AAH32732-06
3-Ethoxy-2,6-difluoroaniline 97%, JRD
3-Ethoxy-2,6-difluoroaniline
Formula: C₈H₉F₂NO
Storage Temperature: Ambient
MDL Number: MFCD11519309
CAS Number: 651734-64-4
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR