4-Methoxy-2-(trifluoromethyl)benzylamine 97%, JRD
|
Danger
|
H32730-03
H32730-06
AAH32730-03EA
149.85
USD
AAH32730-03
AAH32730-06
4-Methoxy-2-(trifluoromethyl)benzylamine 97%, JRD
4-Methoxy-2-(trifluoromethyl)benzylamine
Formula:
C₉H₁₀F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD09025405 CAS Number: 916420-84-3 UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...