3-(3-Methyl-5-(trifluoromethoxy)phenyl)propionic acid 97%, JRD

Warning

H32721-MD H32721-03
AAH32721-03EA 165.39 USD
AAH32721-03 AAH32721-MD
3-(3-Methyl-5-(trifluoromethoxy)phenyl)propionic acid 97%, JRD
3-(3-Methyl-5-(trifluoromethoxy)phenyl)propionic acid
Formula: C₁₁H₁₁F₃O₃
MW: 248.20
Storage Temperature: Ambient
MDL Number: MFCD09025376
CAS Number: 916420-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR