2-Methyl-3-(trifluoromethyl)anisole 97%, JRD

Warning

H32706-03 H32706-MD
AAH32706-03EA 172.05 USD
AAH32706-03 AAH32706-MD
2-Methyl-3-(trifluoromethyl)anisole 97%, JRD
2-Methyl-3-(trifluoromethyl)anisole
Formula: C₉H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD09832284
CAS Number: 1017778-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR