1-Boc-DL-nipecotic acid ethyl ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 1-Boc-3-piperidinecarboxylate
, Ethyl 1-Boc-DL-nipecotate
H32704-06
H32704-03
AAH32704-03EA
51.35
USD
AAH32704-03
AAH32704-06
1-Boc-DL-nipecotic acid ethyl ester 97%
1-Boc-DL-nipecotic acid ethyl ester
Formula:
C₁₃H₂₃NO₄ MW: 257.33 g/mol Boiling Pt: 95 °C/0.5 mm Melting Pt: 31…35 °C Storage Temperature: Ambient |
MDL Number:
MFCD04116274 CAS Number: 130250-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...