3-Ethoxy-2,6-difluorobenzoyl chloride 97%, JRD

Danger

H32700-06 H32700-03
AAH32700-06EA 148.74 USD
AAH32700-06 AAH32700-03
3-Ethoxy-2,6-difluorobenzoyl chloride 97%, JRD
3-Ethoxy-2,6-difluorobenzoyl chloride
Formula: C₉H₇ClF₂O₂
MW: 220.60 g/mol
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD11519369
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR