3-Bromo-5-fluorobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H32692-06 H32692-03
AAH32692-06EA 386.99 USD
AAH32692-06 AAH32692-03
3-Bromo-5-fluorobenzoic acid 97%
3-Bromo-5-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD01569540
CAS Number: 176548-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR