2-(Difluoromethoxy)-4-fluorobenzyl bromide 97%, JRD

Danger

H32676-MD H32676-03
AAH32676-MDEA 84.27 USD
AAH32676-MD AAH32676-03
2-(Difluoromethoxy)-4-fluorobenzyl bromide 97%, JRD
2-(Difluoromethoxy)-4-fluorobenzyl bromide
Formula: C₈H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09832363
CAS Number: 1017779-42-8
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR