2-Methoxy-5-(trifluoromethoxy)benzyl alcohol 98+%
|
Warning
|
H32670-03
H32670-MD
AAH32670-MDEA
44.72
USD
AAH32670-MD
AAH32670-03
2-Methoxy-5-(trifluoromethoxy)benzyl alcohol 98+%
2-Methoxy-5-(trifluoromethoxy)benzyl alcohol
Formula:
C₉H₉F₃O₃ MW: 222.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115960 CAS Number: 562840-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...