Ethyl-1H-indazole-5-carboxylate 95%

Synonyms: Indazole-5-carboxylic acid ethyl esterEthyl Indazole-5-carboxylateEthyl 1H-indazole-5-carboxylate

H32649-MD H32649-03
AAH32649-MDEA 102.38 USD
AAH32649-MD AAH32649-03
Ethyl-1H-indazole-5-carboxylate 95%
Ethyl 1H-indazole-5-carboxylate
Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD03426220
CAS Number: 192944-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR