4-Chloro-3-fluorocinnamic acid 95%

Warning

H32636-03 H32636-09
AAH32636-09EA 220.89 USD
AAH32636-09 AAH32636-03
4-Chloro-3-fluorocinnamic acid 95%
4-Chloro-3-fluorocinnamic acid
Formula: C₉H₆ClFO₂
MW: 200.6 g/mol
Melting Pt: 241…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00143294
CAS Number: 202982-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR