3',5'-Difluoro-2'-hydroxyacetophenone 97%

Warning

Synonyms: 1-(3,5-Difluoro-2-hydroxyphenyl)ethanone

H32624-03 H32624-09
AAH32624-03EA 47.41 USD
AAH32624-03 AAH32624-09
3',5'-Difluoro-2'-hydroxyacetophenone 97%
3',5'-Difluoro-2'-hydroxyacetophenone
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Melting Pt: 105…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00042476
CAS Number: 140675-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR