Acetyl chloride 1 M in dichloromethane

Danger

H32616-AP H32616-AE
AAH32616-APEA 105.06 USD
AAH32616-AP AAH32616-AE
Acetyl chloride 1 M in dichloromethane
Acetyl chloride
Formula: C₂H₃ClO
MW: 78.5 g/mol
Density: 1.294
Storage Temperature: Ambient
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 2922
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR