4-Methoxy-2-(trifluoromethyl)benzamide 97%, JRD

Warning

H32608-03 H32608-06
AAH32608-06EA 386.28 USD
AAH32608-06 AAH32608-03
4-Methoxy-2-(trifluoromethyl)benzamide 97%, JRD
4-Methoxy-2-(trifluoromethyl)benzamide
Formula: C₉H₈F₃NO₂
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD09025404
CAS Number: 916420-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR