5-Nitro-o-anisic acid 96%

Warning

Synonyms: 2-Methoxy-5-nitrobenzoic acid

H32563-03 H32563-06
AAH32563-06EA 213.21 USD
AAH32563-06 AAH32563-03
5-Nitro-o-anisic acid 96%
5-Nitro-o-anisic acid
Formula: C₈H₇NO₅
MW: 197.15 g/mol
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00527902
CAS Number: 40751-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR